Skip to Content
Merck
All Photos(1)

Documents

203195

Sigma-Aldrich

Protein A, Biotin Conjugate

This Protein A, Biotin Conjugate is validated for use in Solid Phase Immunoassay.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116133
NACRES:
NA.41

Quality Level

form

liquid

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

ambient

storage temp.

−20°C

General description

Native protein from Straphylococcus Aureus, conjugated to biotin. Recognizes the Fc portions of immunoglobulins of various species.
Native protein A conjugated to biotin.

Application

Solid Phase Immunoassay (1:10,000)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 10 mM PBS, 1% BSA, pH 7.6.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

A single band at MW 50,000 by SDS-PAGE

Other Notes

The protein A: biotin molar ratio is determined on a lot-specific basis. Binding strength is as follows: strongly reactive, guinea pig, human IgG1, IgG2, and IgG4, and rabbit; weakly reactive: bovine, human IgA, IgM, and IgE, and mouse; nonreactive: goat, horse, human IgD and IgG3, rat, and sheep. o-phenylenediamine, 1.0 mg/ml in citrate buffer, pH 4.5, 0.1% urea peroxide at 25°C, was used as substrate for testing biotin-Protein A and avidin peroxidase functionality. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service