Skip to Content
Merck
All Photos(1)

Key Documents

5.08738

Sigma-Aldrich

Palmostatin B

InSolution, ≥95%, 50 mM in DMSO, APT1 inhibitor

Synonym(s):

InSolution APT1 Inhibitor, palmostatin B, APT1 Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
€94.90

€94.90


Estimated to ship on28 May 2025Details


Request a Bulk Order

Select a Size

Change View
2 MG
€94.90

About This Item

Empirical Formula (Hill Notation):
C23H36O4
Molecular Weight:
376.53
UNSPSC Code:
12352200
NACRES:
NA.77

€94.90


Estimated to ship on28 May 2025Details


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

General description

A cell-permeable, beta-lactone acyl protein thioesterase 1 (APT1) inhibitor (IC50 = 0.67 µM, in an enzymatic assay) that is shown to specifically block Ras depalmitoylation, without affecting Ras acylation, in MDCK cells, both in vitro and in vivo. It induces a marked redistribution of NRas to endomembranes (1 µM) without notable cytotoxicity, and is shown to elicit a loss of the precise steady-state localization of palmitoylated Ras proteins in the same cell line. At 50 µM, this inhibitor displays a partial phenotypic reversion in oncogenic HRasG12V-transformed fibroblasts. Furthermore, it demonstrates selectivity for APT1 over phospholipase A1, A2, Cβ and D. It′s inhibitory effect is demonstrated to be consistent with that of APT1 downregulation by siRNA.

Biochem/physiol Actions

Primary Target
APT1

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

A 50 mM (2 mg/106.23 µL) sterile-filtered solution of APT1 Inhibitor, palmostatin B (Cat. No. 178501). in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C). Aliquots are stable for up to 3 months at -20°C.

Other Notes

Dekker, F. and Hedberg, C. 2011. Bioorg. Med. Chem. Lett.19, 1376.

Dekker, F., et al. 2010. Nat. Chem. Biol.6, 449.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frank J Dekker et al.
Bioorganic & medicinal chemistry, 19(4), 1376-1380 (2010-12-07)
The H- and N-Ras GTPases are prominent examples of proteins, whose localizations and signalling capacities are regulated by reversible palmitoylations and depalmitoylations. Recently, the novel small molecule inhibitor palmostatin B has been described to inhibit Ras depalmitoylation and to revert
Frank J Dekker et al.
Nature chemical biology, 6(6), 449-456 (2010-04-27)
Cycles of depalmitoylation and repalmitoylation critically control the steady-state localization and function of various peripheral membrane proteins, such as Ras proto-oncogene products. Interference with acylation using small molecules is a strategy to modulate cellular localization--and thereby unregulated signaling--caused by palmitoylated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service