Skip to Content
Merck
All Photos(2)

Key Documents

441251

Sigma-Aldrich

α2-Macroglobulin, Human Plasma

A multifunctional, secreted glycoprotein that acts as a broad range irreversible protease inhibitor that forms a "trap" around most proteases.

Synonym(s):

α₂M, α₂MG

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€187.00
10 MG
€1,120.00

€187.00


Please contact Customer Service for Availability


Select a Size

Change View
1 MG
€187.00
10 MG
€1,120.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

€187.00


Please contact Customer Service for Availability

Quality Level

Assay

≥95% (SDS-PAGE)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

aqueous buffer: soluble

shipped in

ambient

storage temp.

−20°C

General description

A multifunctional, secreted glycoprotein that acts as a broad range irreversible protease inhibitor that forms a "trap" around most proteases. α2M is so large a molecule that it tends to remain intravascular. Localized on the surface of peripheral blood lymphocytes. Multifunctional activities include promoting growth of mammalian cells in culture, stimulating regeneration of lymphocytes in irradiated mice, transporting zinc, and acting as proteinase inhibitor that controls the clotting and fibrinolytic system.

Biochem/physiol Actions

Cell permeable: no
Primary Target
broad range protease
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 30 mM sodium phosphate buffer, with glycine, pH 7.0.

Other Notes

Bonner, J.C., et al. 1995. J. Biol. Chem. 270, 16236.
Lysiak, J.J., et al. 1995. J. Biol. Chem.270, 21919.
Boisset, N., et al. 1992. J. Struct. Biol.108, 221.
Barrett, A.J. 1980. Methods Enzymol.80, 737.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Li et al.
International journal of oncology, 57(1), 213-222 (2020-05-08)
Advanced osteoradionecrosis (ORN) is one of the most serious complications in patients with head and neck cancer, resulting in poor prognosis. Numerous studies have therefore focused on the pathogenesis and interventions of ORN early stage. The present study aimed to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service