Skip to Content
ID
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
epT.I.P.S.
®
G Racks, Biopur
®
Products
Product Category
pipette tips (8)
Color
Manufacturer
Color
Package Size
Sterility
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
EP0030075056
Eppendorf
®
epT.I.P.S.
®
G Racks, Biopur
®
,
size (20-300 μL), L (55 mm), orange, pack of 480 ea (5 trays of 96 tips)
Expand
View Pricing
EP0030075153
Eppendorf
®
epT.I.P.S.
®
G Racks, Biopur
®
,
size (0.5-10 mL), L (165 mm), turquoise, pack of 120 ea (5 trays of 24 tips)
Expand
View Pricing
EP0030075013
Eppendorf
®
epT.I.P.S.
®
G Racks, Biopur
®
,
size (2-20 μL), L (40 mm), medium gray, pack of 480 ea (5 trays of 96 tips)
Expand
View Pricing
EP0030075110
Eppendorf
®
epT.I.P.S.
®
G Racks, Biopur
®
,
size (0.5-2.5 mL), L (115 mm), red, pack of 240 ea (5 trays of 48 tips)
Expand
View Pricing
EP0030075030
Eppendorf
®
epT.I.P.S.
®
G Racks, Biopur
®
,
size (2-200 μL), L (53 mm), yellow, pack of 480 ea (5 trays of 96 tips)
Expand
View Pricing
EP0030075072
Eppendorf
®
epT.I.P.S.
®
G Racks, Biopur
®
,
size (50-1000 μL), L (71 mm), blue
Expand
View Pricing
EP0030075161
Eppendorf
®
epT.I.P.S.
®
G Racks, Biopur
®
,
size (50-1250 μL), L (103 mm), dark green, pack of 480 ea (5 trays of 96 tips)
Expand
View Pricing
EP0030075099
Eppendorf
®
epT.I.P.S.
®
G Racks, Biopur
®
,
size (50-1250 μL), L (76 mm), green, pack of 480 ea (5 trays of 96 tips)
Expand
View Pricing