Key Documents
EP0030075099
Eppendorf® epT.I.P.S.® G Racks, Biopur®
size (50-1250 μL), L (76 mm), green, pack of 480 ea (5 trays of 96 tips)
About This Item
UNSPSC Code:
41121600
volume:
50—1250 μL
sterility:
sterile
feature:
DNase free
packaging:
pack of 480 ea (5 trays of 96 tips)
manufacturer/tradename:
Eppendorf® 0030075099
Pricing and availability is not currently available.
material
green (Tray)
sterility
sterile
feature
DNase free
packaging
pack of 480 ea (5 trays of 96 tips)
manufacturer/tradename
Eppendorf® 0030075099
L
(76 mm)
maximum volume
1.25 mL
size
(50-1250 μL)
volume
50—1250 μL
color
green
General description
  • Eppendorf® epT.I.P.S.® G Racks, Biopur® offer the highest biological purity grade, Guaranteed PCR clean, sterile, pyrogen-, ATP- and DNA-free.
  • For use in medical, pharmaceutical and food industry, molecular biology and cell technology.
  • Each production lot inspected continuously by independent laboratory.
  • Packed in racks with 96 tips.
Legal Information
Biopur is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)