Skip to Content
Merck
All Photos(2)

Key Documents

V900339

Sigma-Aldrich

Cytosine β-D-arabinofuranoside

Vetec, reagent grade, 90%

Synonym(s):

(β-D-Arabinofuranosyl)cytosine, Ara-C, Arabinocytidine, Arabinosylcytosine, Cytarabine, Cytosine arabinoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N3O5
CAS Number:
Molecular Weight:
243.22
Beilstein:
89175
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

Assay

90%

storage temp.

2-8°C

SMILES string

NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](CO)[C@@H](O)[C@@H]2O

InChI

1S/C9H13N3O5/c10-5-1-2-12(9(16)11-5)8-7(15)6(14)4(3-13)17-8/h1-2,4,6-8,13-15H,3H2,(H2,10,11,16)/t4-,6-,7+,8-/m1/s1

InChI key

UHDGCWIWMRVCDJ-CCXZUQQUSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Ara-C incorporates into DNA and inhibits DNA replication by forming cleavage complexes with topoisomerase I resulting in DNA fragmentation; does not inhibit RNA synthesis. Anti-leukemia agent.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Low et al.
Internal medicine journal, 43(3), 294-297 (2012-07-05)
Although induction chemotherapy comprising high-dose cytarabine (HiDAC) in combination with idarubicin and etoposide or 'ICE' for adult acute myeloid leukaemia (AML) produces a complete remission rate of nearly 80%, gastrointestinal toxicity is significant. Omission of etoposide may produce similar clinical
Bhupender S Chhikara et al.
Expert opinion on drug delivery, 7(12), 1399-1414 (2010-10-23)
Cytarabine is a polar nucleoside drug used for the treatment of myeloid leukemia and non-Hodgkin's lymphoma. The drug has a short plasma half-life, low stability and limited bioavailability. Overdosing of patients with continuous infusions may lead to side effects. Thus
Jorge E Romaguera et al.
British journal of haematology, 150(2), 200-208 (2010-06-10)
Mantle cell lymphoma (MCL) has a poor overall survival after treatment with conventional chemotherapy. Intense chemoimmunotherapy without consolidation stem cell transplantation is a potential therapeutic option. We report on a prospective Phase II study with rituximab in combination with fractionated
Bob Löwenberg
Blood, 121(1), 26-28 (2013-01-05)
High-dose cytarabine applied during remission induction or as consolidation after attainment of a complete remission has become an established element in the treatment of adults with acute myeloid leukemia. Recent evidence has challenged the need for these exceptionally high-dose levels
Jorge Cortes et al.
The Lancet. Oncology, 14(4), 354-362 (2013-03-05)
Tosedostat is a novel oral aminopeptidase inhibitor with clinical activity in a previous phase 1-2 study in elderly patients with relapsed or refractory acute myeloid leukaemia (AML). We aimed to compare two dosing regimens of tosedostat. In this randomised phase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service