Skip to Content
Merck
All Photos(1)

Documents

SCJ4600032

Sigma-Aldrich

CF 594 aminooxy

suitable for fluorescence

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

form

solid

Quality Level

ε (extinction coefficient)

115000 at 593 nm

absorbance ratio

A280/593 nm 0.08

fluorescence

λex 532 or 561 nm (Flow cytometry laser line)
λex 593 nm; λem 614 nm
λex 594 nm (Microscopy laser line)

suitability

suitable for fluorescence

storage temp.

−20°C

General description

CF594 is a deep red fluorescent dye spectrally similar to Alexa Fluor 594 and Texas Red® dye. When conjugated to proteins, CF594 is significantly brighter than Alexa Fluor 594 due to its high quantum yield and exceptional water solubility. CF594 also has excellent photostability, making it ideal for demanding applications such as confocal microscopy and single molecule imaging. These properties make CF594 the best deep-red dye for labeling proteins and nucleic acids. The dye is particularly useful in combination with our blue fluorescent CF350, green fluorescent CF488A and far red CF647 for multi-color imaging.

Features and Benefits

1. Yields the brightest antibody conjugates among spectrally similar dyes; 2. Extremely photostable; 3. Highly water-soluble

Preparation Note

Protect from light

Legal Information

This product is distributed by Sigma-Aldrich Co. under the authorization of Biotium, Inc. This product is covered by one or more US patents and corresponding patent claims outside the US patents or pending applications owned or licensed by Biotium, Inc. including without limitation: 12/334,387; 12/607,915; 12/699,778; 12/850,578; 61/454,484. In consideration of the purchase price paid by the buyer, the buyer is hereby granted a limited, non-exclusive, non-transferable license to use only the purchased amount of the product solely for the buyer′s own internal research in a manner consistent with the accompanying product literature. Except as expressly granted herein, the sale of this product does not grant to or convey upon the buyer any license, expressly, by implication or estoppel, under any patent right or other intellectual property right of Biotium, Inc. Buyer shall not resell or transfer this product to any third party, or use the product for any commercial purposes, including without limitation, any diagnostic, therapeutic or prophylactic uses. This product is for research use only. Any other uses, including diagnostic uses, require a separate license from Biotium, Inc. For information on purchasing a license to use this product for purposes other than research, contact Biotium, Inc., 3159 Corporate Place, Hayward, CA 94545, Tel: (510) 265-1027. Fax: (510) 265-1352. Email: btinfo@biotium.com.
ALEXA FLUOR is a trademark of Life Technologies
CF is a trademark of Biotium, Inc.
Texas Red is a registered trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service