Skip to Content
Merck
All Photos(1)

Documents

EHU033441

Sigma-Aldrich

MISSION® esiRNA

targeting human XPC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CGTTTCAATAAAGGGGTCCATGAGGACACACACAAGGTTCACCTTCTCTGCCTGCTAGCAAATGGCTTCTATCGAAATAACATCTGCAGCCAGCCAGATCTGCATGCTATTGGCCTGTCCATCATCCCAGCCCGCTTTACCAGAGTGCTGCCTCGAGATGTGGACACCTACTACCTCTCAAACCTGGTGAAGTGGTTCATTGGAACATTTACAGTTAATGCAGAACTTTCAGCCAGTGAACAAGATAACCTGCAGACTACATTGGAAAGGAGATTTGCTATTTACTCTGCTCGAGATGATGAGGAATTGGTCCATATATTCTTACTGATTCTCCGGGCTCTGCAGCTCTTGACCCGGCTGGTATTGTCTCTACAGCCAATTCCTCTGAAGTCAGCAACAGCAAAGGGAAAGA

Ensembl | human accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy Budden et al.
BMC cancer, 18(1), 100-100 (2018-01-28)
Melanoma has two key features, an over-representation of UV-induced mutations and resistance to DNA damaging chemotherapy agents. Both of these features may result from dysfunction of the nucleotide excision repair pathway, in particular the DNA damage detection branch, global genome
Jyh-Cheng Chen et al.
Pharmacology, 102(1-2), 91-104 (2018-06-29)
Etoposide (VP16) is a topoisomerase II inhibitor and has been used for the treatment of non-small cell lung cancer (NSCLC). Xeroderma pigmentosum complementation group C (XPC) protein is a DNA damage recognition factor in nucleotide excision repair and involved in
Jyh-Cheng Chen et al.
Toxicology research, 7(6), 1247-1256 (2018-12-18)
Astaxanthin has been demonstrated to exhibit a wide range of beneficial effects that include anti-cancer and anti-inflammatory properties. Xeroderma pigmentosum complementation group C (XPC) protein is an important DNA damage recognition factor in nucleotide excision repair and is involved in
Hiroyuki Niida et al.
Nature communications, 8, 16102-16102 (2017-07-19)
HBO1, a histone acetyl transferase, is a co-activator of DNA pre-replication complex formation. We recently reported that HBO1 is phosphorylated by ATM and/or ATR and binds to DDB2 after ultraviolet irradiation. Here, we show that phosphorylated HBO1 at cyclobutane pyrimidine
Tiantian Cui et al.
Oncotarget, 6(12), 10060-10072 (2015-04-15)
Xeroderma pigmentosum complementation group C (XPC) protein is an important DNA damage recognition factor in nucleotide excision repair. Deletion of XPC is associated with early stages of human lung carcinogenesis, and reduced XPC mRNA levels predict poor patient outcome for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service