Skip to Content
Merck
All Photos(1)

Documents

A3908

Sigma-Aldrich

Acid Fuchsin calcium salt

certified by the Biological Stain Commission, Dye content ≥60 %

Synonym(s):

Acid Magenta, Acid Rubin, Fuchsin S, SS, SS, ST, or SIII, Rubin S

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C20H17N3O9S3Ca
CAS Number:
Molecular Weight:
579.64
MDL number:
UNSPSC Code:
12171500

grade

certified by the Biological Stain Commission

composition

Dye content, ≥60%

SMILES string

[Ca++].Cc1cc(cc(c1N)S([O-])(=O)=O)\C(c2ccc(N)c(c2)S([O-])(=O)=O)=C3\C=CC(=N)C(=C3)S(O)(=O)=O

InChI

1S/C20H19N3O9S3.Ca/c1-10-6-13(9-18(20(10)23)35(30,31)32)19(11-2-4-14(21)16(7-11)33(24,25)26)12-3-5-15(22)17(8-12)34(27,28)29;/h2-9,21H,22-23H2,1H3,(H,24,25,26)(H,27,28,29)(H,30,31,32);/q;+2/p-2/b19-11-,21-14?;

InChI key

SVIISRNRODSEHG-SZRYGYPJSA-L

Looking for similar products? Visit Product Comparison Guide

Suitability

Certified for use in Van Gieson connective tissue stain, Mallory′s connective tissue stain and Altmann′s mitochondrial stain.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis and testing of biological stains--the Biological Stain Commission Procedures.
D P Penney et al.
Biotechnic & histochemistry : official publication of the Biological Stain Commission, 77(5-6), 237-275 (2003-02-05)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service