Skip to Content
Merck
All Photos(1)

Key Documents

78981

Sigma-Aldrich

Phenylpropiolaldehyde

technical, ≥95% (GC)

Synonym(s):

3-Phenyl-2-propynal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C≡CCHO
CAS Number:
Molecular Weight:
130.14
Beilstein:
1099281
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Pricing and availability is not currently available.

grade

technical

Assay

≥95% (GC)

refractive index

n20/D 1.604 (lit.)
n20/D 1.605

bp

118 °C/13 mmHg (lit.)

density

1.064 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)C#Cc1ccccc1

InChI

1S/C9H6O/c10-8-4-7-9-5-2-1-3-6-9/h1-3,5-6,8H

InChI key

IDASOVSVRKONFS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service