580553
(±)-Taxifolin hydrate
An antioxidant flavonoid that scavenges superoxide (1.5 x 10⁵ M⁻¹ s⁻¹) and suppresses the cytotoxicity of superoxide and hydrogen peroxide in Chinese hamster V79 cells.
Synonym(s):
3,3′,4′,5,7-Pentahydroxydihydroflavone Hydrate, Dihydroquercetin Hydrate, 3,3′,4′,5,7-Pentahydroxyflavanone hydrate, Dihydroquercetin hydrate
About This Item
Recommended Products
Quality Level
Assay
≥95% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
white to off-white
solubility
ethanol: 20 mg/mL
shipped in
ambient
storage temp.
2-8°C
SMILES string
O.O[C@@H]1[C@H](Oc2cc(O)cc(O)c2C1=O)c3ccc(O)c(O)c3
InChI
1S/C15H12O7.H2O/c16-7-4-10(19)12-11(5-7)22-15(14(21)13(12)20)6-1-2-8(17)9(18)3-6;/h1-5,14-19,21H;1H2/t14-,15+;/m0./s1
InChI key
DNQPGSVMBPSRIR-LDXVYITESA-N
Looking for similar products? Visit Product Comparison Guide
General description
Biochem/physiol Actions
An antioxidant flavonoid that scavenges superoxide
Packaging
Warning
Other Notes
Bors, W., et al. 1995. Free Radic. Biol. Med. 19, 45.
Nakayama, T., et al. 1994. Cancer Res. 54, 1991s.
Nakayama, T., et al. 1993. Biochem. Pharmacol. 45, 265.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service