Skip to Content
Merck
All Photos(1)

Key Documents

436526

Sigma-Aldrich

N-(Hydroxymethyl)methacrylamide solution

technical grade, 52-55 wt. % in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CONHCH2OH
CAS Number:
Molecular Weight:
115.13
EC Number:
MDL number:
UNSPSC Code:
12162002

Pricing and availability is not currently available.

grade

technical grade

contains

10-20 ppm monomethyl ether hydroquinone as inhibitor

concentration

52-55 wt. % in H2O

impurities

<6% methacrylamide, < 2% formaldehyde.

refractive index

n20/D 1.43

bp

~100 °C

mp

−37 °C

density

1.1 g/mL at 25 °C

SMILES string

CC(=C)C(=O)NCO

Packaging

Packaged in glass bottles

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service