About This Item
Recommended Products
Assay
97%
bp
246 °C/755 mmHg (lit.)
mp
73-74 °C (lit.)
SMILES string
CCS(=O)(=O)CC
InChI
1S/C4H10O2S/c1-3-7(5,6)4-2/h3-4H2,1-2H3
InChI key
MBDUIEKYVPVZJH-UHFFFAOYSA-N
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service