Skip to Content
Merck
All Photos(2)

Key Documents

235962

Sigma-Aldrich

Trichloromethanesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CSO2Cl
CAS Number:
Molecular Weight:
217.89
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

chunks

mp

137-140 °C (lit.)

SMILES string

ClC(Cl)(Cl)S(Cl)(=O)=O

InChI

1S/CCl4O2S/c2-1(3,4)8(5,6)7

InChI key

ZCPSWAFANXCCOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trichloromethanesulfonyl chloride is an efficient free radical chlorinating agent. It reacts with pent-4-enylcobaloximes in inert solvent under tugsten lamp irradiation to yield 2-(β,β,β- trichloroethyl)sulfolanes. It also reacts with trimethylsilyl enol ethers of acetophenones in the presence of a ruthenium (II) phosphine complex to yield 1-aryl-3,3-dichloropropen-1-one and α-chloroacetophenones.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Reactions of trichloromethanesulfonyl chloride and carbon tetrachloride with silyl enol ethers catalyzed by a ruthenium (II) phosphine complex.
Kamigata N, et al.
Journal of Organometallic Chemistry, 552(1), 9-43 (1998)
TRICHLOROMETHANESULFONYL CHLORIDE AS A SELECTIVE CHLORINATING AGENT1.
Huyser ES.
Journal of the American Chemical Society, 82(19), 5246-5247 (1960)
Homolytic displacement at saturated carbon. Part 9. The reactions of trichloromethanesulfonyl chloride with pent-4-enylcobaloximes and with olefins. A novel route to (trichloroethyl) sulfolanes via an SHi mechanism.
Ashcroft MR, et al.
The Journal of Organic Chemistry, 49(10), 1751-1761 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service