HPLC Analysis of Chlorthalidone Enantiomers on Astec® CHIROBIOTIC® T
![HPLC Analysis of Chlorthalidone Enantiomers on Astec® CHIROBIOTIC® T application for HPLC](/deepweb/assets/sigmaaldrich/product/images/652/527/b8d9b885-1d4b-4697-976c-4a909498481a/800/b8d9b885-1d4b-4697-976c-4a909498481a.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
[A] 10 mM ammonium acetate, pH 4.1; [B] methanol; (80:20, A:B)
flow rate
0.9 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
5 μL
sample
2 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC