HPLC Analysis of Catechols and Resorcinols on Ascentis® RP-Amide
Materials
analytical column
CONDITIONS
column
Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 μm particles (565324-U)
mobile phase
[A] 20 mM phosphoric acid (pH 2.0 unadjusted): [B] acetonitrile; (75:25, A:B)
flow rate
1.5 mL/min
column temp.
30 °C
detector
UV, 270 nm
injection
25 μL
sample
as indicated in 20 mM phosphoric acid (pH 2.0)
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany