Merck
  • The hydrolysis of epoxides catalyzed by inorganic ammonium salts in water: kinetic evidence for hydrogen bond catalysis.
The hydrolysis of epoxides catalyzed by inorganic ammonium salts in water: kinetic evidence for hydrogen bond catalysis.
Physical chemistry chemical physics : PCCP (2017-12-21)
B Nozière, F Fache, A Maxut, B Fenet, A Baudouin, L Fine, C Ferronato
ABSTRACT
Naturally-occurring inorganic ammonium ions have been recently reported as efficient catalysts for some organic reactions in water, which contributes to the understanding of the chemistry in some natural environments (soils, seawater, atmospheric aerosols, …) and biological systems, and is also potentially interesting for green chemistry as many of their salts are cheap and non-toxic. In this work, the effect of NH
Sigma-Aldrich
1-Amino-2-butanol, AldrichCPR
Pricing and availability is not currently available.
Sigma-Aldrich
2-Amino-1-butanol, 97%
Pricing and availability is not currently available.