Merck
  • The effect of coumaryl alcohol incorporation on the structure and composition of lignin dehydrogenation polymers.
The effect of coumaryl alcohol incorporation on the structure and composition of lignin dehydrogenation polymers.
Biotechnology for biofuels (2017-12-08)
Anne E Harman-Ware, Renee M Happs, Brian H Davison, Mark F Davis
ABSTRACT
Lignin dehydrogenation polymers (DHPs) are polymers generated from phenolic precursors for the purpose of studying lignin structure and polymerization processes. Here, DHPs were synthesized using a Zutropfverfahren method with horseradish peroxidase and three lignin monomers, sinapyl (S), coumaryl (H), and coniferyl (G) alcohols, in the presence of hydrogen peroxide. The H monomer was reacted with G and a 1:1 molar mixture of S:G monomers at H molar compositions of 0, 5, 10, and 20 mol% to study how the presence of the H monomer affected the structure and composition of the recovered polymers. At low H concentrations, solid-state NMR spectra suggest that the H and G monomers interact to form G:H polymers that have a lower average molecular weight than the solely G-based polymer or the G:H polymer produced at higher H concentrations. Solid-state NMR and pyrolysis-MBMS analyses suggest that at higher H concentrations, the H monomer primarily self-polymerizes to produce clusters of H-based polymer that are segregated from clusters of G- or S:G-based polymers. Thioacidolysis generally showed higher recoveries of thioethylated products from S:G or S:G:H polymers made with higher H content, indicating an increase in the linear ether linkages. Overall, the experimental results support theoretical predictions for the reactivity and structural influences of the H monomer on the formation of lignin-like polymers.
Sigma-Aldrich
Sinapyl alcohol, technical grade, 80%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mg
Please contact Customer Service for Availability
HUF 89,200.00
Sigma-Aldrich
Allyl methyl sulfone, 96%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
HUF 66,800.00
Sigma-Aldrich
Coniferyl alcohol, 98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 mg
Please contact Customer Service for Availability
HUF 48,800.00
1 g
Please contact Customer Service for Availability
HUF 151,000.00