Merck
  • Exosomes from human mesenchymal stem cells conduct aerobic metabolism in term and preterm newborn infants.
Exosomes from human mesenchymal stem cells conduct aerobic metabolism in term and preterm newborn infants.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2015-12-15)
Isabella Panfoli, Silvia Ravera, Marina Podestà, Claudia Cossu, Laura Santucci, Martina Bartolucci, Maurizio Bruschi, Daniela Calzia, Federica Sabatini, Matteo Bruschettini, Luca Antonio Ramenghi, Olga Romantsik, Danilo Marimpietri, Vito Pistoia, Gianmarco Ghiggeri, Francesco Frassoni, Giovanni Candiano
ABSTRACT
Exosomes are secreted nanovesicles that are able to transfer RNA and proteins to target cells. The emerging role of mesenchymal stem cell (MSC) exosomes as promoters of aerobic ATP synthesis restoration in damaged cells, prompted us to assess whether they contain an extramitochondrial aerobic respiration capacity. Exosomes were isolated from culture medium of human MSCs from umbilical cord of ≥37-wk-old newborns or between 28- to 30-wk-old newborns (i.e.,term or preterm infants). Characterization of samples was conducted by cytofluorometry. Oxidative phosphorylation capacity was assessed by Western blot analysis, oximetry, and luminometric and fluorometric analyses. MSC exosomes express functional respiratory complexes I, IV, and V, consuming oxygen. ATP synthesis was only detectable in exosomes from term newborns, suggestive of a specific mechanism that is not completed at an early gestational age. Activities are outward facing and comparable to those detected in mitochondria isolated from term MSCs. MSC exosomes display an unsuspected aerobic respiratory ability independent of whole mitochondria. This may be relevant for their ability to rescue cell bioenergetics. The differential oxidative metabolism of pretermvs.term exosomes sheds new light on the preterm newborn's clinical vulnerability. A reduced ability to repair damaged tissue and an increased capability to cope with anoxic environment for preterm infants can be envisaged.-Panfoli, I., Ravera, S., Podestà, M., Cossu, C., Santucci, L., Bartolucci, M., Bruschi, M., Calzia, D., Sabatini, F., Bruschettini, M., Ramenghi, L. A., Romantsik, O., Marimpietri, D., Pistoia, V., Ghiggeri, G., Frassoni, F., Candiano, G. Exosomes from human mesenchymal stem cells conduct aerobic metabolism in term and preterm newborn infants.
Sigma-Aldrich
Anti-ATP5B antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Ouabain, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.