Merck
  • Nicotine exploits a COPI-mediated process for chaperone-mediated up-regulation of its receptors.
Nicotine exploits a COPI-mediated process for chaperone-mediated up-regulation of its receptors.
The Journal of general physiology (2014-01-01)
Brandon J Henderson, Rahul Srinivasan, Weston A Nichols, Crystal N Dilworth, Diana F Gutierrez, Elisha D W Mackey, Sheri McKinney, Ryan M Drenan, Christopher I Richards, Henry A Lester
ABSTRACT
Chronic exposure to nicotine up-regulates high sensitivity nicotinic acetylcholine receptors (nAChRs) in the brain. This up-regulation partially underlies addiction and may also contribute to protection against Parkinson's disease. nAChRs containing the α6 subunit (α6* nAChRs) are expressed in neurons in several brain regions, but comparatively little is known about the effect of chronic nicotine on these nAChRs. We report here that nicotine up-regulates α6* nAChRs in several mouse brain regions (substantia nigra pars compacta, ventral tegmental area, medial habenula, and superior colliculus) and in neuroblastoma 2a cells. We present evidence that a coat protein complex I (COPI)-mediated process mediates this up-regulation of α6* or α4* nAChRs but does not participate in basal trafficking. We show that α6β2β3 nAChR up-regulation is prevented by mutating a putative COPI-binding motif in the β3 subunit or by inhibiting COPI. Similarly, a COPI-dependent process is required for up-regulation of α4β2 nAChRs by chronic nicotine but not for basal trafficking. Mutation of the putative COPI-binding motif or inhibition of COPI also results in reduced normalized Förster resonance energy transfer between α6β2β3 nAChRs and εCOP subunits. The discovery that nicotine exploits a COPI-dependent process to chaperone high sensitivity nAChRs is novel and suggests that this may be a common mechanism in the up-regulation of nAChRs in response to chronic nicotine.
Product Description
USP
Nicotine bitartrate dihydrate, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.