Merck
TOF-SIMS with argon gas cluster ion beams: a comparison with C60+.
Analytical chemistry (2011-04-06)
Sadia Rabbani, Andrew M Barber, John S Fletcher, Nicholas P Lockyer, John C Vickerman
ABSTRACT
Time-of-flight secondary ion mass spectrometry (TOF-SIMS) is an established technique for the characterization of solid sample surfaces. The introduction of polyatomic ion beams, such as C(60), has provided the associated ability to perform molecular depth-profiling and 3D molecular imaging. However, not all samples perform equally under C(60) bombardment, and it is probably naïve to think that there will be an ion beam that will be applicable in all situations. It is therefore important to explore the potential of other candidates. A systematic study of the suitability of argon gas cluster ion beams (Ar-GCIBs) of general composition Ar(n)(+), where n = 60-3000, as primary particles in TOF-SIMS analysis has been performed. We have assessed the potential of the Ar-GCIBs for molecular depth-profiling in terms of damage accumulation and sputter rate and also as analysis beams where spectral quality and secondary ion yields are considered. We present results with direct comparison with C(60) ions on the same sample in the same instrument on polymer, polymer additive, and biomolecular samples, including lipids and small peptides. Large argon clusters show reduced damage accumulation compared with C(60) with an approximately constant sputter rate as a function of Ar cluster size. Further, on some samples, large argon clusters produce changes in the mass spectra indicative of a more gentle ejection mechanism. However, there also appears to be a reduction in the ionization of secondary species as the size of the Ar cluster increases.
Sigma-Aldrich
Angiotensin III trifluoroacetate salt hydrate, ≥98% (HPCE)
Pricing and availability is not currently available.