Merck
  • Synthesis and biological evaluation of 4-hydroxy-4-(4-methoxyphenyl)-substituted proline and pyrrolidin-2-ylacetic acid derivatives as GABA uptake inhibitors.
Synthesis and biological evaluation of 4-hydroxy-4-(4-methoxyphenyl)-substituted proline and pyrrolidin-2-ylacetic acid derivatives as GABA uptake inhibitors.
Bioorganic & medicinal chemistry (2012-12-19)
Xueqing Zhao, Jörg Pabel, Georg C Höfner, Klaus T Wanner
ABSTRACT
A series of enantiomerically pure 4-hydroxy-4-(4-methoxyphenyl)-substituted proline and pyrrolidin-2-ylacetic acid derivatives have been synthesized starting from the respective N-protected 4-hydroxy derivatives via oxidation to the corresponding 4-oxo compounds, subsequent addition of organometallic reagents, final hydrolysis and deprotection. The major diastereoisomers obtained by the addition of the Grignard reagents were found to have opposite stereoconfigurations depending on whether cerium trichloride was present or absent as an additive. The final compounds were evaluated for their capability to inhibit the GABA transport proteins GAT1 and GAT3. 4-Hydroxyproline derivatives substituted with a tris(4-methoxyphenyl)methyloxyethyl residue at the nitrogen and a 4-methoxyphenyl group in 4-position showed, with the exception of the (2R,4R)-diastereomer, an improved inhibition at GAT3 compared to the derivatives missing the 4-methoxyphenyl group in 4-position. This may imply that an appropriate lipophilic group at the C-4 position of the proline moiety is beneficial for potent inhibition at GAT3.
Sigma-Aldrich
Pyrrolidine, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Please contact Customer Service for Availability
₪1,086.00
50 μg
Please contact Customer Service for Availability
₪1,939.00
Sigma-Aldrich
Pyrrolidine, FG
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Please contact Customer Service for Availability
₪1,086.00
50 μg
Please contact Customer Service for Availability
₪1,939.00
Sigma-Aldrich
Pyrrolidine, ≥99.0%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Please contact Customer Service for Availability
₪1,086.00
50 μg
Please contact Customer Service for Availability
₪1,939.00
Sigma-Aldrich
Pyrrolidine, ≥99.5%, purified by redistillation
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 μg
Please contact Customer Service for Availability
₪1,086.00
50 μg
Please contact Customer Service for Availability
₪1,939.00