Merck
  • Chemical and technological delivery systems for idebenone: a review of literature production.
Chemical and technological delivery systems for idebenone: a review of literature production.
Expert opinion on drug delivery (2012-10-23)
Claudia Carbone, Rosario Pignatello, Teresa Musumeci, Giovanni Puglisi
ABSTRACT
Idebenone (IDE) is an antioxidant compound, structurally related to coenzyme Q10. Its therapeutic potential is growing in different application areas, as demonstrated by the number of experimental works and patents produced in very recent years. Cyclodextrin inclusion complexes, liposomes, microemulsions, prodrugs, polymeric and lipid nanoparticles have been explored to achieve different goals, such as topical administration, brain targeting or increasing the bioavailability of this highly lipophilic drug. This review summarizes the results of works published in the last 20 years for the delivery and targeting of this drug. A direct comparison of the different carrier systems is not easy and could not even be significant, due to the large variables existing among them. However, the different forms of delivery can help increase idebenone solubility, stability and biochemical activity. Further studies will be developed in order to improve the controlled release and targeting of idebenone.
Sigma-Aldrich
Idebenone, ≥98% (HPLC)
Pricing and availability is not currently available.
Supelco
Idebenone, analytical standard
Pricing and availability is not currently available.