Merck
  • Stability considerations of aspartame in the direct analysis of artificial sweeteners in water samples using high-performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS).
Stability considerations of aspartame in the direct analysis of artificial sweeteners in water samples using high-performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS).
Chemosphere (2012-04-17)
Jean-Daniel Berset, Nicole Ochsenbein
ABSTRACT
A HPLC-MS/MS method is presented for the simultaneous determination of frequently used artificial sweeteners (ASs) and the main metabolite of aspartame (ASP), diketopiperazine (DKP), in environmental water samples using the direct-injection (DI) technique, thereby achieving limits of quantification (LOQ) of 10 ng L(-1). For a reliable quantification of ASP pH should be adjusted to 4.3 to prevent formation of the metabolite. Acesulfame (ACE), saccharin (SAC), cyclamate (CYC) and sucralose (SUC) were ubiquitously found in water samples. Highest concentrations up to 61 μg L(-1) of ACE were found in wastewater effluents, followed by surface water with concentrations up to 7 μg L(-1), lakes up to 600 ng L(-1) and groundwater and tap water up to 70 ng L(-1). The metabolite DKP was only detected in wastewater up to 200 ng L(-1) and at low detection frequencies.
Product Description
Supelco
Aspartame, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Asp-Phe methyl ester, ≥98%
Pricing and availability is not currently available.
Aspartame, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.