Merck
Small GTP-binding proteins.
Physiological reviews (2001-01-12)
Y Takai, T Sasaki, T Matozaki
ABSTRACT
Small GTP-binding proteins (G proteins) exist in eukaryotes from yeast to human and constitute a superfamily consisting of more than 100 members. This superfamily is structurally classified into at least five families: the Ras, Rho, Rab, Sar1/Arf, and Ran families. They regulate a wide variety of cell functions as biological timers (biotimers) that initiate and terminate specific cell functions and determine the periods of time for the continuation of the specific cell functions. They furthermore play key roles in not only temporal but also spatial determination of specific cell functions. The Ras family regulates gene expression, the Rho family regulates cytoskeletal reorganization and gene expression, the Rab and Sar1/Arf families regulate vesicle trafficking, and the Ran family regulates nucleocytoplasmic transport and microtubule organization. Many upstream regulators and downstream effectors of small G proteins have been isolated, and their modes of activation and action have gradually been elucidated. Cascades and cross-talks of small G proteins have also been clarified. In this review, functions of small G proteins and their modes of activation and action are described.
Sigma-Aldrich
Ral B human, recombinant, expressed in E. coli, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
Rap1A human, buffered aqueous glycerol solution, recombinant, expressed in E. coli, GST-tagged
Pricing and availability is not currently available.