Merck
  • Generation of human metabolites of 7-ethoxycoumarin by bacterial cytochrome P450 BM3.
Generation of human metabolites of 7-ethoxycoumarin by bacterial cytochrome P450 BM3.
Drug metabolism and disposition: the biological fate of chemicals (2008-08-02)
Dong-Hyun Kim, Keon-Hee Kim, Dae-Hwan Kim, Kwang-Hyeon Liu, Heung-Chae Jung, Jae-Gu Pan, Chul-Ho Yun
ABSTRACT
Recently, wild-type and mutant forms of bacterial cytochrome P450 BM3 (CYP102A1) have been found to metabolize various drugs through reactions similar to those catalyzed by human cytochromes P450 (P450s). Therefore, it has been suggested that CYP102A1 may be used to produce large quantities of the metabolites of human P450-catalyzed reactions. In this report, we show that the oxidation of 7-ethoxycoumarin, a typical human P450 substrate, is catalyzed by both wild-type and mutant forms of CYP102A1. Two major products were produced as a result of O-deethylation and 3-hydroxylation reactions. These results demonstrate that CYP102A1 mutants catalyze the same reactions as human P450s. High noncompetitive intermolecular kinetic deuterium isotope effects were observed for 7-ethoxycoumarin O-deethylation in the CYP102A1 system. These results suggest that there is a common mechanism for the oxidation reactions catalyzed by both the bacterial CYP102A1 and human P450 enzymes.
Sigma-Aldrich
7-Ethoxycoumarin
Pricing and availability is not currently available.