Merck
  • One-step rapid determination and purification of puerarin from Radix puerariae by n-octylamine-modified poly(methacrylate-co-ethylene dimethacrylate) monolith.
One-step rapid determination and purification of puerarin from Radix puerariae by n-octylamine-modified poly(methacrylate-co-ethylene dimethacrylate) monolith.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2008-07-22)
Yong-Qin Lv, Tian-Wei Tan, Man-Yi Wang, Jan-Christer Janson
ABSTRACT
n-Octylamine-modified poly(methacrylate-co-ethylene dimethacrylate) monoliths were prepared for rapid screening, determination and one-step purification of puerarin from Radix puerariae (a crude extract of the root of Pueraria lobata). The modified monolith showed a specific surface area of 17.8 m(2) g(-1), an average pore size of 0.76 microm and a total porosity of 60.8%. Fast separation of R. puerariae crude extract was achieved within 5 min at a flow velocity of 722 cm h(-1) resulting in a puerarin purity of 97%, with a recovery of 85%. This demonstrates the potential of n-octylamine-modified poly(methacrylate-co-ethylene dimethacrylate) monolith for the rapid analysis and separation of isoflavonoids. Preparative scale sample loading (12 mg in 2 mL) resulted in a purity of 95%, and a recovery of about 69%. HPLC, FTIR, MS and (1)H NMR spectroscopy were used for the characterization and quantification of puerarin in isolated fraction.
Sigma-Aldrich
Octylamine, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Octylamine, 99%