Merck
Enantioselective Synthesis of Azamerone.
Journal of the American Chemical Society (2019-02-02)
Matthew L Landry, Grace M McKenna, Noah Z Burns
ABSTRACT
A concise and selective synthesis of the dichlorinated meroterpenoid azamerone is described. The paucity of tactics for the synthesis of natural-product-relevant chiral organochlorides motivated the development of unique strategies for accessing these motifs in enantioenriched forms. The route features a novel enantioselective chloroetherification reaction, a Pd-catalyzed cross-coupling between a quinone diazide and a boronic hemiester, and a late-stage tetrazine [4+2]-cycloaddition/oxidation cascade.
Sigma-Aldrich
SPhos Pd G3, 97%
Pricing and availability is not currently available.