Merck
  • Perfluorooctanesulfonic acid (PFOS) and perfluorobutanesulfonic acid (PFBS) impaired reproduction and altered offspring physiological functions in Caenorhabditis elegans.
Perfluorooctanesulfonic acid (PFOS) and perfluorobutanesulfonic acid (PFBS) impaired reproduction and altered offspring physiological functions in Caenorhabditis elegans.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2020-08-25)
Yiren Yue, Sida Li, Zhuojia Qian, Renalison Farias Pereira, Jonghwa Lee, Jeffery J Doherty, Zhenyu Zhang, Ye Peng, John M Clark, Alicia R Timme-Laragy, Yeonhwa Park, Yiren Yue, Sida Li, Zhuojia Qian, Renalison Farias Pereira, Jonghwa Lee, Jeffery J Doherty, Zhenyu Zhang, Ye Peng, John M Clark, Alicia R Timme-Laragy, Yeonhwa Park
ABSTRACT
Perfluorobutanesulfonic acid (PFBS), a shorter chain Per- and polyfluoroalkyl substances (PFASs) cognate of perfluorooctanesulfonic acid (PFOS), has been used as replacement for the toxic surfactant PFOS. However, emerging evidences suggest safety concerns for PFBS and its effect on reproductive health is still understudied. Therefore, the current work aimed to investigate the effect of PFBS, in comparison to PFOS, on reproductive health using Caenorhabditis elegans as an in vivo animal model. PFOS (≥10 μM) and PFBS (≥1000 μM) significantly impaired the reproduction capacity of C. elegans, represented as reduced brood size (total egg number) and progeny number (hatched offspring number), without affecting the hatchability. Additionally, the preconception exposure of PFOS and PFBS significantly altered the embryonic nutrient loading and composition, which further led to abnormalities in growth rate, body size and locomotive activity in F1 offspring. Though the effective exposure concentration of PFBS was approximately 100 times higher than PFOS, the internal concentration of PFBS was lower than that of PFOS to produce the similar effects of PFOS. In conclusion, PFOS and PFBS significantly impaired the reproductive capacities in C. elegans and the preconception exposure of these two compounds can lead to offspring physiological dysfunctions.
Sigma-Aldrich
Nonafluorobutane-1-sulfonic acid, 97%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
HUF 22,620.00
25 g
Please contact Customer Service for Availability
HUF 84,250.00