Skip to Content
Merck
Anti-ZC3H14 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-ZC3H14 antibody produced in rabbit

Anti-ZC3H14 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-FLJ11806, Anti-NY-REN-37, Anti-UKp68, Anti-zinc finger CCCH-type containing 14, Anti-MGC26892, Anti-Zinc finger CCCH-type containing 14

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique