![Amoxicillin Related Compound B](/deepweb/assets/sigmaaldrich/product/images/521/243/60b9eeb0-1b46-4f4e-9074-8eef2ecc2a92/640/60b9eeb0-1b46-4f4e-9074-8eef2ecc2a92.jpg)
Amoxicillin Related Compound B
![Amoxicillin Related Compound B](/deepweb/assets/sigmaaldrich/product/images/521/243/60b9eeb0-1b46-4f4e-9074-8eef2ecc2a92/640/60b9eeb0-1b46-4f4e-9074-8eef2ecc2a92.jpg)
Synonym(s):
L-Amoxicillin, (2S,5R,6R)-6-[(S)-2-Amino-2-(4-hydroxyphenyl)acetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Empirical Formula (Hill Notation):
C16H19N3O5S
CAS Number:
Molecular Weight:
365.40
Products
Formula Weight
Manufacturer
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing