Skip to Content
Merck
All Photos(1)

Key Documents

O8208

Sigma-Aldrich

Orcinol monohydrate

97%

Synonym(s):

3,5-Dihydroxytoluene, 5-Methylresorcinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3-1,3-(OH)2 · H2O
CAS Number:
Molecular Weight:
142.15
Beilstein:
1071903
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Assay

97%

bp

290 °C (lit.)

mp

56-58 °C (lit.)

SMILES string

O.Cc1cc(O)cc(O)c1

InChI

1S/C7H8O2.H2O/c1-5-2-6(8)4-7(9)3-5;/h2-4,8-9H,1H3;1H2

InChI key

NBKPNAMTHBIMLA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for sugars.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Smadar Shulami et al.
The Journal of biological chemistry, 289(37), 25957-25975 (2014-07-30)
Geobacillus stearothermophilus T-6 produces a single extracellular xylanase (Xyn10A) capable of producing short, decorated xylo-oligosaccharides from the naturally branched polysaccharide, xylan. Gel retardation assays indicated that the master negative regulator, XylR, binds specifically to xylR operators in the promoters of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service