Skip to Content
Merck
All Photos(1)

Documents

MABS2282

Sigma-Aldrich

Anti-SIRPA Antibody, clone MY-1

Synonym(s):

Tyrosine-protein phosphatase non-receptor type substrate 1;SHP substrate 1;SHPS-1;Brain Ig-like molecule with tyrosine-based activation motifs;Bit;CD172 antigen-like family member A;Inhibitory receptor SHPS-1;MyD-1 antigen;Signal-regulatory protein alpha-

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

MY-1, monoclonal

mol wt

calculated mol wt 56.14 kDa
observed mol wt ~95 kDa

purified by

using protein G

species reactivity

mouse

packaging

antibody small pack of 100 μL

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG2aκ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Sirpa(19261)

Related Categories

General description

Tyrosine-protein phosphatase non-receptor type substrate 1 (UniProt: P97797; also known as SHP substrate 1, SHPS-1, Brain Ig-like molecule with tyrosine-based activation motifs, Bit, CD172 antigen-like family member A, Inhibitory receptor SHPS-1, MyD-1 antigen, Signal-regulatory protein alpha-1, Sirp-alpha-1, mSIRP-alpha1, p84, CD172a) is encoded by the Sirpa (also known as Myd1, Ptpns1, Shps1, Sirp) gene (Gene ID: 19261) in murine species. SIRPA is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-31), which is subsequently cleaved off to produce the mature protein that contains an extracellular domain (aa 32-373), a transmembrane domain (aa 374-394), and a cytoplasmic domain (aa 395-511). Its extracellular region contains one Ig-like V-type domain (aa 32-137) and two Ig-like C1 type domains (aa 149-248 and 255-343). It is highly expressed in cerebral cortex, brain, spinal cord, cerebellum, and spleen, and at much lower levels in kidney, thymus, heart, lung, and liver. Within the cerebellum, highly expressed throughout the molecular layer, and in synaptic glomeruli in the granule cell layer. SIRPA serves as an immunoglobulin-like cell surface receptor for CD47, and their interactions is reported to mediate negative regulation of several monocyte/ macrophage functions. CD47 binding prevents maturation of immature dendritic cells and inhibits cytokine production by mature dendritic cells. SIRPA also acts as a docking protein and induces translocation of PTPN6, PTPN11, and other binding partners from the cytosol to the plasma membrane. SIRPA is phosphorylated on tyrosine residues in response to stimulation with EGF, growth hormone, insulin, and platelet derived growth factor. It is also reported to mediate negative regulation of phagocytosis, mast cell activation and dendritic cell activation. CD47 binding prevents maturation of immature dendritic cells and inhibits cytokine production by mature dendritic cells. Four isoforms of SIRPA have been described that are produced by alternative splicing. (Ref.: Sakamoto, M., et al. (2022). Proc. Natl. Acad. Sci. USA. 119(1); e2109923118; Yanagita, T., et al. (2017). JCI Insight. 2(1); e89140).

Specificity

Clone MY-1 is a rat monoclonal antibody that detects Tyrosine-protein phosphatase non-receptor type substrate 1 (SIRPA/CD172a).

Immunogen

Eosinophil-enriched cell fraction from the mouse small intestinal lamina propria.

Application

Quality Control Testing

Evaluated by Western Blotting in Mouse brain tissue extracts.

Western Blotting Analysis: A 1:500 dilution of this antibody detected SIRPA in Mouse brain tissue extracts.

Tested applications


Flow Cytometry Analysis: A representative lot detected SIRPA in Flow Cytometry applications (Verjan Garcia, N., et al. (2011). J Immunol. 187(5):2268-77; Yanagita, T., et al. (2017). JCI Insight. 2(1); e89140).

Immunofluorescence Analysis: A representative lot detected SIRPA in Immunofluorescence applications (Verjan Garcia, N., et al. (2011). J Immunol.;187(5):2268-77; Yanagita, T., et al. (2017). JCI Insight. 2(1):e89140).

Inhibition Assay: A representative lot inhibited the growth of subcutaneously transplanted cancer cells in murine models. (Yanagita, T., et al. (2017). JCI Insight.;2(1):e89140; Sakamoto, M., et al. (2022). Proc Natl Acad Sci USA.;119(1)e2109923118).

Immunoprecipitation Analysis: A representative lot immunoprecipitated SIRPA in Immunoprecipitation applications (Verjan Garcia, N., et al. (2011). J Immunol. 187(5):2268-77; Yanagita, T., et al. (2017). JCI Insight. 2(1):e89140).

Western Blotting Analysis: A representative lot detected SIRPA in Western Blotting applications (Verjan Garcia, N., et al. (2011). J Immunol. 187(5):2268-77; Yanagita, T., et al. (2017). JCI Insight. 2(1):e89140).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-SIRPA, clone MY-1, Cat. No. MABS2282, is a rat monoclonal antibody that detects SIRPA/CD172a and is tested for use in Flow Cytometry, Immunofluorescence, Inhibition Assay, Immunoprecipitation, and Western Blotting.

Physical form

Purified rat monoclonal antibody IgG2a in PBS without preservatives.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service