Skip to Content
Merck
All Photos(1)

Documents

341262

Sigma-Aldrich

Factor B, Human

Factor B, Human, is a single-chain glycoprotein present in normal human serum that is cleaved during the activation of complement via the alternative pathway into two fragments of 30 and 63 kDa.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

Assay

≥90% (SDS-PAGE)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

impurities

IgG, IgA, IgM, C3, C4, Factor H, Factor I, or albumin, trace (Ouchterlony)

shipped in

dry ice

storage temp.

−70°C

General description

Factor B, Human, is a single-chain glycoprotein present in normal human serum that is cleaved during the activation of complement via the alternative pathway into two fragments of 30 and 63 kDa.
Native factor B from human serum. Single-chain glycoprotein present in normal human serum at about 200 µg/ml. During activation of complement via the alternative pathway, Factor B is cleaved and thus activated by Factor D into two fragments of M.W. of 30 kDa (Ba) and 63 kDa (Bb). The Bb fragment contains the serine protease enzymatic active site. The activated Bb fragment continues activation via the alternative pathway by cleaving, and thus activating, C3 and C5.
Native factor B from human serum. Single-chain glycoprotein present in normal human serum at about 200 µg/ml. During activation of complement via the alternative pathway, Factor B is cleaved, and thus activated by Factor D into two fragments of M.W. of ~30 kDa (Ba) and ~63 kDa (Bb). The Bb fragment contains the serine protease enzymatic active site. The activated Bb fragment continues activation via the alternative pathway by cleaving, and thus activating, C3 and C5.

Biochem/physiol Actions

≥1,200 Factor BH50 units/mg; ≥50% of Factor B activity in NHS (mg/mg basis)

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, pH 7.2.

Preparation Note

Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Kam, C.M., et al. 1987. J. Biol. Chem. 262, 3444.
Gotze, O. and Muller-Eberhard, H.J. 1971. J. Exp. Med. 134, 90s.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

not combustible

Flash Point(C)

not combustible


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service