Skip to Content
Merck
All Photos(1)

Documents

1.03913

Millipore

KLIGLER agar

for the identification of Gram-negative intestinal bacteria

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

form

medium granules (dehydrated (DCM))

manufacturer/tradename

GranuCult®

technique(s)

microbiological culture: suitable

pH

7.3-7.5 (37 °C, 55 g/L in H2O, after autoclaving)

solubility

55 g/L

bulk density

620 kg/m3

application(s)

clinical
microbiology

storage temp.

15-25°C

suitability

enterobacteriaceae

General description

for the identification of Gram-negative intestinal bacteria

Analysis Note

Appearance (clearness)clearAppearance (colour)redpH-value (25 °C)7.2 - 7.6Growth (Escherichia coli ATCC 25922 (WDCM 00013))good to very goodGrowth (Citrobacter freundii ATCC 8090)good to very goodGrowth (Enterobacter cloacae ATCC 13047 (WDCM 00083))good to very goodGrowth (Shigella flexneri ATCC 12022 (WDCM 00126))good to very goodGrowth (Salmonella typhimurium ATCC 14028 (WDCM 00031))good to very goodGrowth (Salmonella enteritidis ATCC 13076 (WDCM 00030))good to very goodGrowth (Proteus mirabilis ATCC 14153)good to very goodGrowth (Proteus hauseri ATCC 13315)good to very goodButt (Escherichia coli ATCC 25922 (WDCM 00013))yellowButt (Citrobacter freundii ATCC 8090)yellow and blackButt (Enterobacter cloacae ATCC 13047 (WDCM 00083))yellowButt (Shigella flexneri ATCC 12022 (WDCM 00126))yellowButt (Salmonella typhimurium ATCC 14028 (WDCM 00031))yellow and blackButt (Salmonella enteritidis ATCC 13076 (WDCM 00030))yellow and blackButt (Proteus mirabilis ATCC 14153)yellowButt (Proteus hauseri ATCC 13315)yellow and blackSlant surface (Escherichia coli ATCC 25922 (WDCM 00013))yellowSlant surface (Citrobacter freundii ATCC 8090)yellowSlant surface (Enterobacter cloacae ATCC 13047 (WDCM 00083))yellowSlant surface (Shigella flexneri ATCC 12022 (WDCM 00126))redSlant surface (Salmonella typhimurium ATCC 14028 (WDCM 00031))redSlant surface (Salmonella enteritidis ATCC 13076 (WDCM 00030))redSlant surface (Proteus mirabilis ATCC 14153)redSlant surface (Proteus hauseri ATCC 13315)redIncubation: 24 hrs.; 35 °C; aerobic.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xizhong Jing et al.
Molecular biology of the cell, 34(7), br10-br10 (2023-05-03)
The short-chain fatty acid metabolite acetyl-coenzyme A (acetyl-CoA) has emerged as a major signal transducer that can broadly affect cell fate and function, at least partly by influencing acetylation of key proteins. The mechanism by which acetyl-CoA regulates CD4+ T-cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service