Skip to Content
Merck
All Photos(1)

Key Documents

ENAH961CE2D8

Enamine

5-methyl-2-(propan-2-yl)phenyl acetate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
HUF 63,042.00
250 MG
HUF 83,790.00
500 MG
HUF 144,058.00
1 G
HUF 191,482.00
2.5 G
HUF 358,948.00

Vendor SKU: EN300-22486452-100MG

HUF 63,042.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
HUF 63,042.00
250 MG
HUF 83,790.00
500 MG
HUF 144,058.00
1 G
HUF 191,482.00
2.5 G
HUF 358,948.00

About This Item

CAS Number:
Molecular Weight:
192.26
MDL number:

Vendor SKU: EN300-22486452-100MG

HUF 63,042.00


Ships in 14 days - from Aldrich Partner.

Iupac Name

2-isopropyl-5-methylphenyl acetate

Inchi Code

1S/C12H16O2/c1-8(2)11-6-5-9(3)7-12(11)14-10(4)13/h5-8H,1-4H3

InChI key

WFMIUXMJJBBOGJ-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-22486452

MSDS

https://www.enaminestore.com/catalog/EN300-22486452


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service