Skip to Content
Merck
All Photos(1)

Key Documents

ENAH95E7255B

Enamine

4-methyl-1,2,3,4-tetrahydroquinoxalin-6-ol dihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
HUF 176,662.00
100 MG
HUF 254,714.00
250 MG
HUF 355,984.00
500 MG
HUF 551,608.00
1 G
HUF 702,772.00
2.5 G
HUF 1,360,286.00
5 G
HUF 2,005,450.00

Vendor SKU: EN300-27159023-50MG

HUF 176,662.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
HUF 176,662.00
100 MG
HUF 254,714.00
250 MG
HUF 355,984.00
500 MG
HUF 551,608.00
1 G
HUF 702,772.00
2.5 G
HUF 1,360,286.00
5 G
HUF 2,005,450.00

About This Item

CAS Number:
Molecular Weight:
237.13
MDL number:

Vendor SKU: EN300-27159023-50MG

HUF 176,662.00


Ships in 14 days - from Aldrich Partner.

Iupac Name

4-methyl-1,2,3,4-tetrahydroquinoxalin-6-ol dihydrochloride

Inchi Code

1S/C9H12N2O.2ClH/c1-11-5-4-10-8-3-2-7(12)6-9(8)11;;/h2-3,6,10,12H,4-5H2,1H3;2*1H

InChI key

GFSYBCMFXRFHFS-UHFFFAOYSA-N

storage temp.

4

Salt Data

Cl

Purity

95

Country of Origin

UA

Shipping Temperature

Ice Pack

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-27159023

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-27159023


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service