Skip to Content
Merck
All Photos(1)

Key Documents

ENAH11DCFC7B

Enamine

{2-oxaspiro[4.4]nonan-3-yl}methanesulfonyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
HUF 175,180.00
250 MG
HUF 243,352.00
500 MG
HUF 373,768.00
1 G
HUF 474,050.00
2.5 G
HUF 912,722.00
5 G
HUF 1,342,008.00
10 G
HUF 1,981,244.00

Vendor SKU: EN300-368348-100MG

HUF 175,180.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
HUF 175,180.00
250 MG
HUF 243,352.00
500 MG
HUF 373,768.00
1 G
HUF 474,050.00
2.5 G
HUF 912,722.00
5 G
HUF 1,342,008.00
10 G
HUF 1,981,244.00

About This Item

CAS Number:
Molecular Weight:
238.73
MDL number:

Vendor SKU: EN300-368348-100MG

HUF 175,180.00


Ships in 14 days - from Aldrich Partner.

Iupac Name

(2-oxaspiro[4.4]nonan-3-yl)methanesulfonyl chloride

Inchi Code

1S/C9H15ClO3S/c10-14(11,12)6-8-5-9(7-13-8)3-1-2-4-9/h8H,1-7H2

InChI key

ZWYVSLBHRSYHQX-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-368348

MSDS

https://www.enaminestore.com/catalog/EN300-368348


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service