Skip to Content
Merck
All Photos(1)

Key Documents

ENA497373642

Enamine

1-[(ethanesulfonyl)methyl]-4-nitrobenzene

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
HUF 31,426.00
250 MG
HUF 37,354.00
500 MG
HUF 49,210.00
1 G
HUF 58,102.00

Vendor SKU: EN300-248282-100MG

HUF 31,426.00


Availability
Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
HUF 31,426.00
250 MG
HUF 37,354.00
500 MG
HUF 49,210.00
1 G
HUF 58,102.00

About This Item

CAS Number:
Molecular Weight:
229.26
MDL number:

Vendor SKU: EN300-248282-100MG

HUF 31,426.00


Availability
Ships in 14 days - from Aldrich Partner.

Iupac Name

1-[(ethylsulfonyl)methyl]-4-nitrobenzene

Inchi Code

1S/C9H11NO4S/c1-2-15(13,14)7-8-3-5-9(6-4-8)10(11)12/h3-6H,2,7H2,1H3

InChI key

QRLVYWVPKPUBJJ-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-248282

MSDS

https://www.enaminestore.com/catalog/EN300-248282


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service