Skip to Content
Merck
All Photos(1)

Key Documents

ENA372573019

Enamine

2-(pentan-2-yl)pyrrolidine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
HUF 143,070.00
250 MG
HUF 196,916.00
500 MG
HUF 299,668.00
1 G
HUF 379,696.00
2.5 G
HUF 727,472.00
5 G
HUF 1,068,332.00
10 G
HUF 1,576,658.00

Vendor SKU: EN300-112023-100MG

HUF 143,070.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
HUF 143,070.00
250 MG
HUF 196,916.00
500 MG
HUF 299,668.00
1 G
HUF 379,696.00
2.5 G
HUF 727,472.00
5 G
HUF 1,068,332.00
10 G
HUF 1,576,658.00

About This Item

CAS Number:
Molecular Weight:
141.26
MDL number:

Vendor SKU: EN300-112023-100MG

HUF 143,070.00


Ships in 14 days - from Aldrich Partner.

Iupac Name

2-(1-methylbutyl)pyrrolidine

Inchi Code

1S/C9H19N/c1-3-5-8(2)9-6-4-7-10-9/h8-10H,3-7H2,1-2H3

InChI key

IXCHPPILTWOTIC-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-112023

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service