Skip to Content
Merck
All Photos(1)

Key Documents

ENA311759455

Enamine

2-cyclopentylacetaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
HUF 146,984.00
250 MG
HUF 154,888.00
500 MG
HUF 169,708.00
1 G
HUF 181,070.00
2.5 G
HUF 243,314.00
5 G
HUF 340,138.00
10 G
HUF 519,954.00

Vendor SKU: EN300-70446-100MG

HUF 146,984.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
HUF 146,984.00
250 MG
HUF 154,888.00
500 MG
HUF 169,708.00
1 G
HUF 181,070.00
2.5 G
HUF 243,314.00
5 G
HUF 340,138.00
10 G
HUF 519,954.00

About This Item

CAS Number:
Molecular Weight:
112.17
MDL number:

Vendor SKU: EN300-70446-100MG

HUF 146,984.00


Ships in 14 days - from Aldrich Partner.

Iupac Name

cyclopentylacetaldehyde

Inchi Code

1S/C7H12O/c8-6-5-7-3-1-2-4-7/h6-7H,1-5H2

InChI key

CEUXIAUEIGSQSZ-UHFFFAOYSA-N

storage temp.

-10

Purity

95

Country of Origin

UA

Shipping Temperature

Ice Pack

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-70446

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service