Skip to Content
Merck
All Photos(1)

Key Documents

ENA087844930

Enamine

3-{[3-(benzyloxy)phenyl]methylidene}-9-oxo-1H,2H,3H,9H-pyrrolo[2,1-b]quinazoline-6-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
HUF 63,042.00
250 MG
HUF 83,790.00

Vendor SKU: EN300-24288-100MG

HUF 63,042.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
HUF 63,042.00
250 MG
HUF 83,790.00

About This Item

CAS Number:
Molecular Weight:
424.46
MDL number:

Vendor SKU: EN300-24288-100MG

HUF 63,042.00


Ships in 14 days - from Aldrich Partner.

Iupac Name

(3E)-3-[3-(benzyloxy)benzylidene]-9-oxo-1,2,3,9-tetrahydropyrrolo[2,1-b]quinazoline-6-carboxylic acid

Inchi Code

1S/C26H20N2O4/c29-25-22-10-9-20(26(30)31)15-23(22)27-24-19(11-12-28(24)25)13-18-7-4-8-21(14-18)32-16-17-5-2-1-3-6-17/h1-10,13-15H,11-12,16H2,(H,30,31)/b19-13+

InChI key

XQQNKZQUJUNYMB-CPNJWEJPSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-24288

MSDS

https://www.enaminestore.com/catalog/EN300-24288


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service