Skip to Content
Merck
All Photos(1)

Documents

W6365

Sigma-Aldrich

Sodium acetate-1-13C

API for Clinical Studies, 99 atom % 13C

Synonym(s):

Acetic acid-1-13C sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3 13CO2Na
CAS Number:
Molecular Weight:
83.03
Beilstein:
4335203
MDL number:
UNSPSC Code:
12352116

grade

cGMP

isotopic purity

99 atom % 13C

mp

>300 °C (dec.) (lit.)

mass shift

M+1

storage temp.

15-25°C

SMILES string

[Na+].C[13C]([O-])=O

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1/i2+1;

InChI key

VMHLLURERBWHNL-CGOMOMTCSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service