614955
Cyclohexanol-OD
99 atom % D
Synonym(s):
Cyclohexan(ol-d), Cyclohexyl alcohol-OD
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H11OD
Molecular Weight:
101.17
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
isotopic purity
99 atom % D
bp
160-161 °C (lit.)
mp
20-22 °C (lit.)
density
0.957 g/mL at 25 °C
mass shift
M+1
SMILES string
[2H]OC1CCCCC1
InChI
1S/C6H12O/c7-6-4-2-1-3-5-6/h6-7H,1-5H2/i7D
InChI key
HPXRVTGHNJAIIH-WHRKIXHSSA-N
Related Categories
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service