Skip to Content
Merck
All Photos(1)

Key Documents

572381

Sigma-Aldrich

Cerium(IV) oxide-yttria doped

nanopowder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CeO2
Molecular Weight:
172.11
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
Pricing and availability is not currently available.

form

nanopowder

contains

15 mol % yttria as dopant

composition

Ce0.85Y0.15O2

particle size

<100 nm

SMILES string

O=[Ce]=O.O=[Y]O[Y]=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service