Key Documents
57171
Supelco
Disco para SPE ENVI-18 DSK
diam. 47 mm, pk of 24
About This Item
UNSPSC Code:
41115712
agency
suitable for EPA 1613
Quality Level
product line
ENVI
packaging
pk of 24
technique(s)
solid phase extraction (SPE): suitable
diam.
47 mm
application(s)
food and beverages
separation technique
reversed phase
General description
Las equivalentes membranas SPE de ENVI-18 y ENVI-8 empaquetadas como sorbentes de lecho SPE
Mecanismo de Retención: Fase Reversa
Compatibilidad de la Matriz de la muestra: Soluciones Acuosas (agua de bebida)
• Membranas de fibra de vidrio porosa embebidas con partículas de sílica modificadas C18 o C8
• Proporciona caudales más rápidos y exhibe menos obstáculos que los discos de Teflón para la extracción de contaminantes orgánicos de muestras de bebidas acuosas
• Aplicaciones típicas incluyen hidrocarburos aromáticos polinucleares (PAHs), policlorados bifenilos (PCBs), ftalatos, orgánicos semivolátiles, paraquat, diquat, pesticidas y herbicidas
Legal Information
ENVI is a trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Lot/Batch Number
Lo sentimos, en este momento no disponemos de COAs para este producto en línea.
Visite la Librería de documentos
Los clientes también vieron
Artículos
Reversed-phase interaction will retain most molecules with hydrophobic character; it is very useful for extracting analytes that are very diverse in structure within the same sample.
Contenido relacionado
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.
Chromatograms
suitable for GC, application for SPE