509493
SGE Gas Tight Syringes, Luer Lock
2.5MDF-LL-GT, volume 2500 μL, needle size (not included)
Sinónimos:
SYRINGE SGE 2.5ML, LUER LOCK
About This Item
Productos recomendados
material
glass syringe
needle size
(not included)
feature
needle type Luer lock
packaging
pkg of 1 ea
manufacturer/tradename
SGE 008425
volume
2500 μL
Categorías relacionadas
1 of 4
Este artículo | 509477 | 509469 | 509485 |
---|---|---|---|
pkg of 1 ea | pkg of 1 ea | pkg of 1 ea | pkg of 1 ea |
SGE 008425 | SGE 006230 | SGE 005230 | SGE 007230 |
2500 μL | 0.25 mL (250 μL) | 100 μL | 500 μL |
(not included) | (not included) | (not included) | (not included) |
needle type Luer lock | without needle, Fixed Luer Lock | Fixed Luer Lock | Fixed Luer Lock |
General description
- Maximum inertness for liquid or gas samples
- Suitable for manual disperser/diluter applications
- High accuracy of dispensed volume
- Wide variety of terminations
- Less time consuming
- SGE gas tight syringes also improves work flow analysis
related product
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)