Documentos clave
RF451211-EP
Rotor for Eppendorf® MiniSpin® / MiniSpin® Plus
F-45-12-11, non aerosol tight black, to hold, 12 x 1.5/2.0 mL tubes, lid
About This Item
UNSPSC Code:
41103909
feature
lid
to hold 12 x 1.5/2.0 mL tubes
fixed angle rotor
non aerosol tight black
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022668498 (NA)
Eppendorf® 5452720008 (EU)
parameter
12,100 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models MiniSpin/MiniSpin Plus
Categorías relacionadas
General description
Rotors for use with Eppendorf MiniSpin and MiniSpin Plus. Rotors are fixed angle and include quick-Lock lids.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Minispin is a registered trademark of Eppendorf AG