Documentos clave
DWKW242833-A
Wheaton® Dropping Bottle
white bottle, capacity 7 mL, case of 1000
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
white bottle
packaging
pkg of 1 packs
case of 1000
manufacturer/tradename
WHEATON® W242833-A
capacity
7 mL
Application
Opthalmic, IVD Kits, Other
Features and Benefits
  • Bottle made of natural or white LDPE (Low Density Polyethylene
  • White LDPE bottle helps protect UV light sensitive samples
  • Dropper tip caps and tips sold separately
  • Packaged in cases of 100 or 1000
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)