BR36403
BRAND® BLAUBRAND® volumetric flask trapezoidal
capacity 5 mL, polypropylene stopper, joint: ST/NS 7/16
Sinónimos:
BLAUBRAND®, Volumetric flasks
About This Item
Productos recomendados
material
polypropylene stopper
transparent borosilicate glass 3.3 flask
joint
joint: ST/NS7/16
feature
accuracy class A
packaging
pack of 2 ea
manufacturer/tradename
BRAND 36403
capacity
5 mL
1 of 4
Este artículo | BR36401 | BR36408 | BR36402 |
---|---|---|---|
accuracy class A | accuracy class A | accuracy class A (To contain) | accuracy class A |
BRAND 36403 | BRAND 36401 | BRAND 36408 | BRAND 36402 |
5 mL | 1 mL | 50 mL | 2 mL |
joint: ST/NS7/16 | joint ST/NS neck7/16 | joint: ST/NS12/21 | joint: ST/NS7/16 |
pack of 2 ea | pack of 2 ea | pack of 2 ea | pack of 2 ea |
General description
All BLAUBRAND® volumetric flasks are delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer. On demand also available with individual certificate or DAkkS calibration certificate.
Legal Information
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)